Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512170.1 | XP_011510472.1 | 1 - 2 | 99 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011512170.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |

STK17B
Genomic Map