Assay Details
| Gene Symbol: FZD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003468.3 | NP_003459.2 | 2 | 1327 | 82 | |
| GenBank mRNA | AB043702.1 | - | 2 | 1302 | 82 | |
| AK315338.1 | - | 1 | 1176 | 82 | ||
| BC114346.2 | - | 1 | 970 | 82 | ||
| U43318.1 | - | 2 | 1237 | 82 | ||
More Information
Set Membership: |
|

FZD5
Genomic Map