Assay Details
| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712292.3 | XP_006712355.1 | 2 - 3 | 533 | 61 | |
| XM_006712294.1 | XP_006712357.1 | 2 - 3 | 533 | 61 | ||
More Information
Set Membership: |
|

MTERF4
Genomic Map