Assay Details
Gene Symbol: C2orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085437.2 | NP_001078906.2 | 1 - 2 | 677 | 62 | |
XM_011511877.1 | XP_011510179.1 | 2 - 3 | 669 | 62 | ||
GenBank mRNA | AK055735.1 | - | 1 - 2 | 1360 | 62 | |
AK056601.1 | - | 1 - 2 | 677 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000454476.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | ENST00000454476.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000307486.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000388934.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000402775.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000414499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000454476.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | ENST00000454476.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001085437.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_001282921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016336.hg.1 | NM_024861.3 | Refseq |
