Assay Details
Gene Symbol: C2orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC014578.2 | - | 1 - 2 | 61 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
