Assay Details
Gene Symbol: C2orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143959.1 | NP_001137431.1 | 2 - 3 | 228 | 96 | |
NM_001143960.2 | NP_001137432.1 | 2 - 3 | 241 | 96 | ||
XM_017003967.1 | XP_016859456.1 | 2 - 3 | 229 | 96 | ||
GenBank mRNA | BC014578.2 | - | 4 - 5 | 451 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143960.2, XM_017003967.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000398622.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000426997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000432605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000464909.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000488469.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | ENST00000489686.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016455.hg.1 | NM_001143960.2 | Refseq |
