Assay Details
| Gene Symbol: HK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000189.4 | NP_000180.2 | 1 | 522 | 86 | |
| GenBank mRNA | AF086198.1 | - | 1 | 532 | 86 | |
| BM912287.1 | - | 86 | ||||
| CB160837.1 | - | 1 | 86 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_000189.4 transcripts.
Set Membership: |
|

HK2
Genomic Map