Assay Details
| Gene Symbol: FAHD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511282.2 | XP_011509584.1 | 9 | 4133 | 68 | |
| XM_011511283.2 | XP_011509585.1 | 10 | 3510 | 68 | ||
| XM_011511284.2 | XP_011509586.1 | 10 | 3639 | 68 | ||
| XM_011511285.2 | XP_011509587.1 | 10 | 3507 | 68 | ||
| XM_011511286.2 | XP_011509588.1 | 9 | 3393 | 68 | ||
| XM_011511287.2 | XP_011509589.1 | 9 | 3504 | 68 | ||
| XM_011511288.2 | XP_011509590.1 | 9 | 4231 | 68 | ||
| XM_011511289.2 | XP_011509591.1 | 9 | 5300 | 68 | ||
| XM_017004237.1 | XP_016859726.1 | 10 | 4806 | 68 | ||
| XM_017004238.1 | XP_016859727.1 | 10 | 4674 | 68 | ||
| XM_017004240.1 | XP_016859729.1 | 9 | 4671 | 68 | ||
| XM_017004241.1 | XP_016859730.1 | 7 | 4415 | 68 | ||
| XM_017004242.1 | XP_016859731.1 | 7 | 3675 | 68 | ||
| XM_017004245.1 | XP_016859734.1 | 6 | 4252 | 68 | ||
| XM_017004246.1 | XP_016859735.1 | 7 | 4114 | 68 | ||
| XM_017004247.1 | XP_016859736.1 | 6 | 3512 | 68 | ||
| XM_017004248.1 | XP_016859737.1 | 6 | 3623 | 68 | ||
| XM_017004249.1 | XP_016859738.1 | 5 | 3443 | 68 | ||
| GenBank mRNA | AK092980.1 | - | 1 | 1518 | 68 | |
More Information
Set Membership: |
|

FAHD2A
Genomic Map