Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004931.4 | NP_004922.1 | 3 - 4 | 552 | 96 | |
| NM_172101.3 | NP_742099.1 | 3 - 4 | 552 | 96 | ||
| NM_172213.3 | NP_757362.1 | 3 - 4 | 552 | 96 | ||
| XM_011533164.2 | XP_011531466.1 | 3 - 4 | 510 | 96 | ||
| GenBank mRNA | AK303360.1 | - | 3 - 4 | 510 | 96 | |
| AK303595.1 | - | 2 - 3 | 406 | 96 | ||
| BC100911.1 | - | 3 - 4 | 543 | 40003720 | 96 | |
| BC100913.1 | - | 3 - 4 | 543 | 40003724 | 96 | |
| BC100914.1 | - | 3 - 4 | 543 | 40003725 | 96 | |
| BQ004743.1 | - | 3 - 4 | 96 | |||
| M36712.1 | - | 3 - 4 | 538 | 96 | ||
| X13444.1 | - | 3 - 4 | 543 | 96 | ||
| X13445.1 | - | 3 - 4 | 543 | 96 | ||
| X13452.1 | - | 3 - 4 | 507 | 96 | ||
| Y00805.1 | - | 3 - 4 | 538 | 96 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000416057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008766.hg.1 | ENST00000417670.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000331469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000349455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000390655.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393759.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000393761.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | ENST00000431506.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_001178100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_004931.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172101.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013324.hg.1 | NM_172213.3 | Refseq |

CD8B
Genomic Map