Assay Details
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011511772.1 | XP_011510074.1 | 2 - 3 | 195 | 76 | |
XM_011511776.1 | XP_011510078.1 | 2 - 3 | 194 | 76 | ||
XM_017004851.1 | XP_016860340.1 | 2 - 3 | 205 | 76 | ||
XM_017004853.1 | XP_016860342.1 | 2 - 3 | 205 | 76 | ||
XM_017004854.1 | XP_016860343.1 | 2 - 3 | 205 | 76 | ||
GenBank mRNA | AK131289.1 | - | 2 - 3 | 196 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011511776.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
