| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |