Assay Details
| Gene Symbol: BRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018462.4 | NP_060932.2 | 2 - 3 | 252 | 93 | |
| GenBank mRNA | AF161418.1 | - | 2 - 3 | 213 | 92 | |
| AF220047.1 | - | 1 - 2 | 93 | |||
| AY148219.1 | - | 2 - 3 | 252 | 93 | ||
| AY148220.1 | - | 2 - 3 | 252 | 93 | ||
| BC001067.1 | - | 2 - 3 | 228 | 2823162 | 93 | |
| BC007929.1 | - | 2 - 3 | 221 | 3688264 | 93 | |
| BC019303.1 | - | 2 - 3 | 219 | 2823162 | 93 | |
| BC061515.1 | - | 2 - 3 | 226 | 93 | ||
More Information
Set Membership: |
|

BRK1
Genomic Map