Assay Details
Gene Symbol: C3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198562.2 | NP_940964.1 | 3 | 1996 | 108 | |
GenBank mRNA | AK056215.1 | - | 1 | 815 | 108 | |
AK096799.1 | - | 2 | 1740 | 108 | ||
AK125642.1 | - | 3 | 1941 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007154.hg.1 | ENST00000584365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007154.hg.1 | ENST00000584365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007371.hg.1 | ENST00000362676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007371.hg.1 | ENST00000362676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000296327.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000416660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000428985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000442203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000471430.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000472653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000475672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000476129.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000484407.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | ENST00000492794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013918.hg.1 | NM_152672.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007154.hg.1 | ENST00000584365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007154.hg.1 | ENST00000584365.1 | Ensembl |
