| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000464919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000464919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000318887.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000464919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000464919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000485219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
ENST00000486971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008018.hg.1 |
NM_173824.3 |
Refseq |