Assay Details
| Gene Symbol: COL6A4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027898.1 | 14 - 15 | 757 | 104 | ||
| GenBank mRNA | CN285077.1 | - | 12 - 13 | 661 | 103 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000509855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000430317.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000504443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000507516.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000509090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000509855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | ENST00000542114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008816.hg.1 | NR_027898.1 | Refseq |

COL6A4P2
Genomic Map