Assay Details
| Gene Symbol: ST3GAL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323358.1 | NP_001310287.1 | 5 - 6 | 654 | 113 | |
| NM_001323359.1 | NP_001310288.1 | 4 - 5 | 554 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323358.1, NM_001323359.1 transcripts.
Set Membership: |
|

ST3GAL6
Genomic Map