Assay Details
| Gene Symbol: EAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC014589.1 | - | 3 - 4 | 408 | 104 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000273668.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000451944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000465664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | ENST00000490477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008536.hg.1 | NM_018456.5 | Refseq |

EAF2
Genomic Map