Assay Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198621.2 | NP_001185550.1 | 1 - 2 | 154 | 74 | |
| NM_194293.2 | NP_919269.2 | 1 - 2 | 144 | 74 | ||
| GenBank mRNA | AJ626899.1 | - | 1 - 2 | 152 | 74 | |
| AJ626900.1 | - | 1 - 2 | 152 | 74 | ||
| AY375160.1 | - | 1 - 2 | 144 | 74 | ||
| BX648565.1 | - | 1 - 2 | 154 | 74 | ||
| CR749430.1 | - | 1 - 2 | 154 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ626899.1, AJ626900.1, AY375160.1, CR749430.1, NM_001198621.2, NM_194293.2 transcripts.
Set Membership: |
|

XIRP1
Genomic Map