Assay Details
| Gene Symbol: KCNMB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126560.1 | 6 - 7 | 710 | 78 | ||
| GenBank mRNA | AY769439.1 | - | 6 - 7 | 657 | 78 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000421498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000425330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000432385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000437488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | ENST00000451742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013249.hg.1 | NR_126561.1 | Refseq |

KCNMB2-AS1
Genomic Map