Assay Details
| Gene Symbol: ADCY5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199642.1 | NP_001186571.1 | 9 - 10 | 1140 | 60 | |
| NM_183357.2 | NP_899200.1 | 9 - 10 | 2115 | 60 | ||
| XM_005247077.3 | XP_005247134.1 | 9 - 10 | 2202 | 60 | ||
| XM_005247078.1 | XP_005247135.1 | 9 - 10 | 1140 | 60 | ||
| XM_006713483.1 | XP_006713546.1 | 9 - 10 | 1136 | 60 | ||
| XM_006713484.1 | XP_006713547.1 | 9 - 10 | 1141 | 60 | ||
| XM_011512359.2 | XP_011510661.1 | 9 - 10 | 2000 | 60 | ||
| XM_011512360.2 | XP_011510662.1 | 9 - 10 | 1197 | 60 | ||
| XM_011512361.1 | XP_011510663.1 | 9 - 10 | 1104 | 60 | ||
| XM_017005638.1 | XP_016861127.1 | 9 - 10 | 1032 | 60 | ||
| XM_017005639.1 | XP_016861128.1 | 9 - 10 | 1032 | 60 | ||
| XM_017005640.1 | XP_016861129.1 | 9 - 10 | 1128 | 60 | ||
| GenBank mRNA | AF497517.1 | - | 9 - 10 | 1016 | 60 | |
| AK093840.1 | - | 10 - 11 | 1104 | 60 | ||
| AK124691.1 | - | 10 - 11 | 1552 | 60 | ||
| AK294565.1 | - | 9 - 10 | 1308 | 60 | ||
| AK303070.1 | - | 9 - 10 | 1141 | 60 | ||
| BX647739.1 | - | 6 - 7 | 617 | 60 | ||
More Information
Set Membership: |
|

ADCY5
Genomic Map