Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271650.1 | NP_001258579.1 | 7 - 8 | 1242 | 72 | |
| NM_022461.4 | NP_071906.1 | 6 - 7 | 1178 | 72 | ||
| XM_005265392.2 | XP_005265449.1 | 7 - 8 | 1265 | 72 | ||
| XM_011534009.2 | XP_011532311.1 | 6 - 7 | 718 | 72 | ||
| GenBank mRNA | AK025592.1 | - | 6 - 7 | 1025 | 72 | |
| AK290637.1 | - | 6 - 7 | 897 | 72 | ||
| AY151386.1 | - | 5 - 6 | 646 | 72 | ||
| BM546093.1 | - | 5 - 6 | 514 | 72 | ||
| BX648471.1 | - | 7 - 8 | 906 | 72 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map