Assay Details
| Gene Symbol: GK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039547.2 | NP_001034636.1 | 15 - 16 | 1593 | 96 | |
| NR_033289.1 | 16 - 17 | 2676 | 96 | |||
| XM_011512624.2 | XP_011510926.1 | 15 - 16 | 1391 | 96 | ||
| XM_017006089.1 | XP_016861578.1 | 16 - 17 | 1656 | 96 | ||
| XM_017006090.1 | XP_016861579.1 | 14 - 15 | 1504 | 96 | ||
| GenBank mRNA | AK090901.1 | - | 6 - 7 | 642 | 96 | |
| AK127641.1 | - | 15 - 16 | 1561 | 96 | ||
| BC032470.1 | - | 16 - 17 | 2664 | 5217372 | 96 | |
| BX648359.1 | - | 15 - 16 | 1573 | 96 | ||
| BX648681.1 | - | 16 - 17 | 2613 | 96 | ||
More Information
Set Membership: |
|

GK5
Genomic Map