Assay Details
| Gene Symbol: LINC01206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104146.1 | 8 | 6407 | 88 | ||
| GenBank mRNA | BX641067.1 | - | 1 | 3137 | 88 | |
| CR749396.1 | - | 1 | 2327 | 88 | ||
More Information
Set Membership: |
|

LINC01206
Genomic Map