| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000425008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000609871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013354.hg.1 |
ENST00000625709.1 |
Ensembl |