Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011520238.2 | XP_011518540.1 | 1 - 2 | 787 | 91 | |
XM_011520239.2 | XP_011518541.1 | 1 - 2 | 787 | 91 | ||
XM_011520240.2 | XP_011518542.1 | 1 - 2 | 787 | 91 | ||
XM_011520243.2 | XP_011518545.1 | 1 - 2 | 787 | 91 | ||
XM_011520244.2 | XP_011518546.1 | 1 - 2 | 787 | 91 | ||
XM_017018070.1 | XP_016873559.1 | 1 - 2 | 787 | 91 | ||
XM_017018071.1 | XP_016873560.1 | 1 - 2 | 787 | 91 | ||
XM_017018079.1 | XP_016873568.1 | 1 - 2 | 787 | 91 | ||
GenBank mRNA | BX951676.1 | - | 1 - 2 | 205 | 91 | |
DA387604.1 | - | 1 - 2 | 212 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011520238.2, XM_011520239.2, XM_011520240.2, XM_011520243.2, XM_011520244.2, XM_017018070.1, XM_017018071.1, XM_017018079.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000278198.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000527150.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000528697.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000530763.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000533474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000534577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | ENST00000619527.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_001258419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NM_020929.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010580.hg.1 | NR_047674.1 | Refseq |
