Assay Details
| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057749.1 | - | 1 | 1098 | 93 | |
| AK074594.1 | - | 1 | 1113 | 93 | ||
| BC032312.1 | - | 1 | 1069 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBRG1| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057749.1 | - | 1 | 1098 | 93 | |
| AK074594.1 | - | 1 | 1113 | 93 | ||
| BC032312.1 | - | 1 | 1069 | 93 | ||
Set Membership: |
|