Assay Details
| Gene Symbol: LOC283278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833334.1 | - | 1 | 1417 | 73 | |
| AL833346.1 | - | 1 | 1417 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283278| Gene Symbol: LOC283278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833334.1 | - | 1 | 1417 | 73 | |
| AL833346.1 | - | 1 | 1417 | 73 | ||
Set Membership: |
|