Assay Details
| Gene Symbol: TRNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124137.1 | - | 3 | 1611 | 115 | |
| BC005184.1 | - | 3 | 652 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRNT1| Gene Symbol: TRNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124137.1 | - | 3 | 1611 | 115 | |
| BC005184.1 | - | 3 | 652 | 115 | ||
Set Membership: |
|