| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000522832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013967.hg.1 |
ENST00000522832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000515154.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000522832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013967.hg.1 |
ENST00000522832.1 |
Ensembl |