| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000502410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000502410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000502410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000502410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
ENST00000509974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007526.hg.1 |
NR_046827.1 |
Refseq |