Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006454.2 | NP_006445.1 | 6 | 3289 | 94 | |
| XM_011513388.2 | XP_011511690.1 | 7 | 3794 | 94 | ||
| GenBank mRNA | AF082740.1 | - | 6 | 3289 | 94 | |
| AK026392.1 | - | 1 | 1427 | 94 | ||
| BC028340.2 | - | 1 | 94 | |||
More Information
Set Membership: |
|

MXD4
Genomic Map