Assay Details
| Gene Symbol: IL21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104126.1 | 6 | 1500 | 148 | ||
| GenBank mRNA | BC045668.1 | - | 6 | 1500 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL21-AS1| Gene Symbol: IL21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104126.1 | 6 | 1500 | 148 | ||
| GenBank mRNA | BC045668.1 | - | 6 | 1500 | 148 | |
Set Membership: |
|