| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000499583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000517335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000523301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
ENST00000624206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009246.hg.1 |
NR_109890.1 |
Refseq |