Assay Details
| Gene Symbol: C5orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009766.1 | XP_016865255.1 | 3 - 4 | 543 | 90 | |
| GenBank mRNA | AL832176.1 | - | 4 - 5 | 900 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009766.1 transcripts.
Set Membership: |
|

C5orf42
Genomic Map