Assay Details
| Gene Symbol: MTMR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543058.2 | XP_011541360.1 | 5 - 6 | 569 | 86 | |
| XM_011543059.2 | XP_011541361.1 | 5 - 6 | 403 | 86 | ||
More Information
Set Membership: |
|

MTMR2
Genomic Map