Assay Details
| Gene Symbol: MUC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002458.2 | NP_002449.2 | 31 | 6413 | 72 | |
| GenBank mRNA | AF253321.1 | - | 1 | 66 | 72 | |
| U78550.1 | - | 1 | 717 | 72 | ||
| X74370.1 | - | 1 | 1332 | 71 | ||
| X74955.1 | - | 1 | 1488 | 72 | ||
| X74956.1 | - | 1 | 791 | 71 | ||
More Information
Set Membership: |
|

MUC5B
Genomic Map