Assay Details
| Gene Symbol: OTOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277269.1 | NP_001264198.1 | 51 - 52 | 8325 | 59 | |
| NM_001292063.1 | NP_001278992.1 | 52 - 53 | 8289 | 59 | ||
More Information
Set Membership: |
|

OTOG
Genomic Map