Assay Details
| Gene Symbol: ARHGEF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074500.1 | - | 1 | 128 | 99 | |
| BC050031.1 | - | 1 | 318 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050031.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |

ARHGEF17
Genomic Map