Assay Details
| Gene Symbol: CWF19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303404.1 | NP_001290333.1 | 9 - 10 | 1052 | 58 | |
| NM_001303405.1 | NP_001290334.1 | 9 - 10 | 1097 | 58 | ||
| NM_001303406.1 | NP_001290335.1 | 6 - 7 | 786 | 58 | ||
| NM_001303407.1 | NP_001290336.1 | 8 - 9 | 821 | 58 | ||
| NM_018294.5 | NP_060764.3 | 9 - 10 | 1052 | 58 | ||
| GenBank mRNA | AK001860.1 | - | 9 - 10 | 998 | 58 | |
| AK097895.1 | - | 8 - 9 | 820 | 58 | ||
| AK225590.1 | - | 9 - 10 | 1010 | 58 | ||
| AK295303.1 | - | 6 - 7 | 756 | 58 | ||
| AL832515.1 | - | 9 - 10 | 1009 | 58 | ||
| BC008746.2 | - | 9 - 10 | 993 | 3345869 | 58 | |
| KU178692.1 | - | 9 - 10 | 964 | 58 | ||
| KU178693.1 | - | 9 - 10 | 948 | 58 | ||
More Information
Set Membership: |
|

CWF19L1
Genomic Map