Assay Details
| Gene Symbol: LOC387720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127642.1 | - | 1 | 623 | 105 | |
| BC132942.1 | - | 1 | 562 | 105 | ||
| BC132944.1 | - | 1 | 562 | 105 | ||
More Information
Set Membership: |
|

LOC387720
Genomic Map