Assay Details
| Gene Symbol: MARVELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031484.3 | NP_113672.1 | 2 | 82 | ||
| GenBank mRNA | AF325259.1 | - | 2 | 82 | ||
| AI347102.1 | - | 1 | 133 | 82 | ||
| AK025996.1 | - | 2 | 82 | |||
| BC004995.1 | - | 2 | 82 | |||
| BC062544.1 | - | 2 | 82 | |||
More Information
Set Membership: |
|

MARVELD1
Genomic Map