Assay Details
| Gene Symbol: KCNMA1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120655.1 | 1 - 2 | 239 | 116 | ||
| GenBank mRNA | BC030624.1 | - | 1 - 2 | 244 | 116 | |
| BX092192.1 | - | 1 - 2 | 207 | 116 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000426234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000429850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000458661.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000595702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000598613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000600782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000608791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000609102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000611475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | ENST00000617241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008123.hg.1 | NR_120655.1 | Refseq |

KCNMA1-AS1
Genomic Map