| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000559920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000560587.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
ENST00000561122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010872.hg.1 |
NR_015419.2 |
Refseq |