| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000053243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000396495.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
ENST00000562385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006940.hg.1 |
NM_001192.2 |
Refseq |