Assay Details
| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311018.1 | - | 1 - 2 | 148 | 131 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000421226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006885.hg.1 | ENST00000580773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000219204.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000562023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000563234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | ENST00000565794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007992.hg.1 | NM_012106.3 | Refseq |

ARL2BP
Genomic Map