Assay Details
| Gene Symbol: MED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100816.1 | NP_001094286.1 | 2 | 609 | 126 | |
| NM_004270.4 | NP_004261.1 | 2 | 325 | 126 | ||
| GenBank mRNA | AF031383.1 | - | 2 | 283 | 126 | |
| AF104251.1 | - | 1 | 217 | 126 | ||
| AK308393.1 | - | 2 | 290 | 126 | ||
| AV711090.1 | - | 2 | 337 | 126 | ||
| BC005250.1 | - | 2 | 279 | 126 | ||
| BC006968.1 | - | 2 | 279 | 126 | ||
| BT007368.1 | - | 1 | 217 | 126 | ||
| CR457009.1 | - | 1 | 217 | 126 | ||
| CR541696.1 | - | 1 | 217 | 126 | ||
More Information
Set Membership: |
|

MED7
Genomic Map