Assay Details
| Gene Symbol: EBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290360.2 | NP_001277289.1 | 16 | 4721 | 100 | |
| NM_001324101.1 | NP_001311030.1 | 17 | 4790 | 100 | ||
| NM_001324103.1 | NP_001311032.1 | 17 | 4787 | 100 | ||
| NM_001324106.1 | NP_001311035.1 | 17 | 4763 | 100 | ||
| NM_001324107.1 | NP_001311036.1 | 16 | 4697 | 100 | ||
| NM_001324108.1 | NP_001311037.1 | 16 | 4694 | 100 | ||
| NM_001324109.1 | NP_001311038.1 | 16 | 4610 | 100 | ||
| NM_001324111.1 | NP_001311040.1 | 16 | 4816 | 100 | ||
| NM_024007.4 | NP_076870.1 | 16 | 4718 | 100 | ||
| NM_182708.2 | NP_874367.1 | 15 | 4625 | 100 | ||
| XM_017009198.1 | XP_016864687.1 | 16 | 5078 | 100 | ||
| XM_017009201.1 | XP_016864690.1 | 11 | 22205 | 100 | ||
| GenBank mRNA | AK096708.1 | - | 1 | 817 | 100 | |
| BM802733.1 | - | 1 | 226 | 100 | ||
More Information
Set Membership: |
|

EBF1
Genomic Map