Assay Details
| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007172.3 | NP_009103.2 | 8 | 1769 | 101 | |
| NM_153645.2 | NP_705931.1 | 9 | 1595 | 101 | ||
| XM_005261312.1 | XP_005261369.1 | 9 | 1819 | 101 | ||
| XM_005261314.1 | XP_005261371.1 | 9 | 1794 | 101 | ||
| XM_006724103.1 | XP_006724166.1 | 8 | 1439 | 101 | ||
| XM_006724104.1 | XP_006724167.1 | 9 | 1629 | 101 | ||
| XM_011529833.1 | XP_011528135.1 | 9 | 1845 | 101 | ||
| XM_017028537.1 | XP_016884026.1 | 9 | 1759 | 101 | ||
| XM_017028538.1 | XP_016884027.1 | 7 | 1430 | 101 | ||
| GenBank mRNA | AF107840.1 | - | 8 | 1472 | 101 | |
| AF116624.2 | - | 7 | 1324 | 101 | ||
| AK095860.1 | - | 3 | 2436 | 101 | ||
| AK304223.1 | - | 7 | 1428 | 101 | ||
| AK314454.1 | - | 8 | 1628 | 101 | ||
| BC016055.1 | - | 9 | 1704 | 4856954 | 101 | |
| BC028125.1 | - | 9 | 1595 | 5166132 | 101 | |
| BC070133.1 | - | 8 | 1629 | 6463796 | 101 | |
| CR456533.1 | - | 7 | 1323 | 101 | ||
More Information
Set Membership: |
|

NUP50
Genomic Map