Assay Details
| Gene Symbol: MYO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248719.3 | XP_005248776.1 | 28 - 29 | 3401 | 123 | |
| XM_005248720.3 | XP_005248777.1 | 28 - 29 | 3401 | 123 | ||
| XM_005248721.3 | XP_005248778.1 | 28 - 29 | 3401 | 123 | ||
More Information
Set Membership: |
|

MYO6
Genomic Map