| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000612407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000612407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000395064.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000612407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000612407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000612775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
ENST00000621915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014112.hg.1 |
NM_152735.3 |
Refseq |